Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106960.1 | NP_001100430.1 | 13 | 1629 | ||
XM_008773161.1 | XP_008771383.1 | 13 | 1711 |