Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256887.2 | XP_006256949.1 | 12 | 1262 | ||
XM_217612.5 | XP_217612.5 | 11 | 1385 |