Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005669.4 | NP_005660.4 | 4, 5 | 673 | ||
GenBank mRNA | AK294496.1 | - | 3, 4 | 659 | ||
AK295436.1 | - | 3, 4 | 425 | |||
AK298268.1 | - | 3, 4 | 408 | |||
AK299902.1 | - | 5, 6 | 937 | |||
AK300048.1 | - | 4, 5 | 491 | |||
AK310729.1 | - | 4, 5 | 1037 | |||
AK312522.1 | - | 4, 5 | 578 | |||
AY562243.1 | - | 4, 5 | 529 | |||
BC000232.1 | - | 4, 5 | 554 | 3350749 | ||
BC065926.1 | - | 4, 5 | 532 | 6164621 | ||
BX648005.1 | - | 4, 5 | 654 | |||
M74090.1 | - | 4, 5 | 555 |