Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181770.1 | NP_861435.1 | 1 | 127 | ||
GenBank mRNA | AY288429.1 | - | 1 | 127 |
Set Membership: |
G-protein coupled receptor |