Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257554.2 | XP_006257616.2 | 5 | 842 | ||
XM_006257555.2 | XP_006257617.1 | 5 | 865 | |||
XM_229133.8 | XP_229133.5 | 5 | 842 |