Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr112l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773667.1 | XP_008771889.1 | 2 | 409 |
Set Membership: |
G-protein coupled receptor |