Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106987.1 | NP_001100457.1 | 3, 4 | 313 | ||
XM_006245952.2 | XP_006246014.1 | 4, 5 | 489 |