Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177515.1 | NP_001170986.1 | 12 | 1425 | ||
NM_021160.2 | NP_066983.1 | 14 | 1298 | |||
NR_033488.1 | 14 | 1549 | ||||
NR_033489.1 | 12 | 1174 | ||||
GenBank mRNA | AF193047.2 | - | 14 | 1247 | ||
AK001207.1 | - | 9 | 745 | |||
AK023194.1 | - | 14 | 1249 | |||
AK025398.1 | - | 1 | 1299 | |||
AK055255.1 | - | 14 | 1549 | |||
AK289469.1 | - | 14 | 1294 | |||
AK297712.1 | - | 12 | 1422 | |||
BC031839.1 | - | 14 | 1253 | 4931559 |