Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277157.1 | NP_001264086.1 | 9 | 2074 | ||
XM_008767581.1 | XP_008765803.1 | 8 | 2294 |