Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107000.1 | NP_001100470.1 | 15 | 2385 | ||
XM_008767683.1 | XP_008765905.1 | 13 | 2075 |