Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002904.5 | NP_002895.3 | 10 | 1189 | ||
XM_006715205.3 | XP_006715268.1 | 10 | 1058 | |||
XM_011514913.2 | XP_011513215.1 | 10 | 1129 | |||
XM_017011299.1 | XP_016866788.1 | 10 | 1114 | |||
GenBank mRNA | AK300717.1 | - | 10 | 972 | ||
AK300898.1 | - | 9 | 1312 | |||
AK302652.1 | - | 10 | 1176 | |||
AK303152.1 | - | 8 | 1484 | |||
BC025235.1 | - | 10 | 1040 | 4876045 | ||
BC050617.2 | - | 10 | 1056 | 6168937 | ||
JQ313904.1 | - | 9 | 985 | |||
L03411.1 | - | 10 | 1071 | |||
X16105.1 | - | 7 | 855 |