Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107005.2 | NP_001100475.2 | 2 | 199 | ||
XM_006246487.2 | XP_006246549.1 | 2 | 357 |