Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166538.1 | NP_001160010.1 | 3 | 443 | ||
NM_007161.3 | NP_009092.3 | 4 | 536 | |||
NM_205837.2 | NP_995309.2 | 4 | 622 | |||
NM_205838.2 | NP_995310.2 | 4 | 559 | |||
NM_205839.2 | NP_995311.2 | 5 | 652 | |||
NM_205840.2 | NP_995312.2 | 3 | 492 | |||
NR_029461.1 | 3 | 536 | ||||
NR_029462.1 | 3 | 491 | ||||
XM_006715206.3 | XP_006715269.1 | 5 | 665 | |||
XM_006715209.3 | XP_006715272.1 | 4 | 568 | |||
XM_006715210.3 | XP_006715273.1 | 4 | 623 | |||
XM_011514914.2 | XP_011513216.1 | 5 | 646 | |||
GenBank mRNA | AF000424.1 | - | 4 | 622 | ||
AF000425.1 | - | 5 | 630 | |||
AF000426.1 | - | 3 | 491 | |||
BC103855.1 | - | 3 | 427 | 40003000 | ||
BC103856.1 | - | 3 | 427 | 40003004 | ||
BX647293.1 | - | 2 | 1718 |