Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168357.1 | NP_001161829.1 | 8 | 868 | ||
NM_005084.3 | NP_005075.3 | 8 | 959 | |||
XM_005249408.4 | XP_005249465.1 | 8 | 943 | |||
GenBank mRNA | AK290381.1 | - | 8 | 918 | ||
AK294471.1 | - | 7 | 741 | |||
AK297066.1 | - | 5 | 535 | |||
BC038452.1 | - | 8 | 868 | 5216829 | ||
U20157.1 | - | 8 | 851 | |||
U24577.1 | - | 8 | 906 |