Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039017.1 | NP_001034106.1 | 5 | 2023 | ||
XM_006246502.2 | XP_006246564.1 | 5 | 2082 | |||
XM_006246503.2 | XP_006246565.1 | 3 | 1881 | |||
XM_008767821.1 | XP_008766043.1 | 5 | 2061 | |||
XM_008767822.1 | XP_008766044.1 | 5 | 2366 | |||
XM_008767823.1 | XP_008766045.1 | 5 | 2369 | |||
GenBank mRNA | BC099105.1 | - | 5 | 2023 | 7460369 |