Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100968.2 | NP_001094438.2 | 12, 13 | 1628 | ||
GenBank mRNA | BC161799.1 | - | 12, 13 | 1628 | 7100780 | |
DQ445463.1 | - | 12, 13 | 1640 | |||
FQ221529.1 | - | 1, 2 | 39 |