Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134645.1 | NP_001128117.1 | 1 | 474 | ||
XM_006246701.2 | XP_006246763.1 | 2 | 848 | |||
XM_006246702.2 | XP_006246764.1 | 1 | 1122 | |||
GenBank mRNA | EF532342.1 | - | 1 | 474 | ||
EF532343.1 | - | 1 | 526 | |||
EF532344.1 | - | 1 | 533 | |||
EF532345.1 | - | 1 | 526 | |||
EF532346.1 | - | 1 | 533 | |||
EF532347.1 | - | 1 | 533 | |||
EF532348.1 | - | 1 | 533 | |||
EF532349.1 | - | 1 | 533 | |||
EF532350.1 | - | 1 | 533 |