Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134645.1 | NP_001128117.1 | 2 | 510 | ||
XM_006246701.2 | XP_006246763.1 | 3 | 884 | |||
XM_006246702.2 | XP_006246764.1 | 2 | 1158 | |||
XM_008767825.1 | XP_008766047.1 | 2 | 218 | |||
GenBank mRNA | EF532342.1 | - | 2 | 510 | ||
EF532343.1 | - | 2 | 562 | |||
EF532344.1 | - | 2 | 569 | |||
EF532345.1 | - | 2 | 562 | |||
EF532346.1 | - | 2 | 569 | |||
EF532347.1 | - | 2 | 569 | |||
EF532348.1 | - | 2 | 569 | |||
EF532349.1 | - | 2 | 569 | |||
EF532350.1 | - | 2 | 569 |