Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107016.1 | NP_001100486.1 | 3, 4 | 382 | ||
GenBank mRNA | FQ233279.1 | - | 3, 4 | 254 |