Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246858.2 | XP_006246920.1 | 21, 22 | 6936 | ||
XM_006246859.2 | XP_006246921.1 | 21, 22 | 6933 | |||
XM_006246860.2 | XP_006246922.1 | 21, 22 | 6921 | |||
XM_006246861.2 | XP_006246923.1 | 21, 22 | 6918 | |||
XM_006246862.2 | XP_006246924.1 | 21, 22 | 3721 | |||
XM_006246864.2 | XP_006246926.1 | 22, 23 | 3756 | |||
XM_008767914.1 | XP_008766136.1 | 21, 22 | 3771 | |||
XM_008767915.1 | XP_008766137.1 | 21, 22 | 3831 | |||
XM_008767916.1 | XP_008766138.1 | 22, 23 | 3469 | |||
XM_220693.9 | XP_220693.6 | 21, 22 | 3704 |