Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107024.1 | NP_001100494.1 | 11 | 1230 | ||
XM_006246905.2 | XP_006246967.1 | 12 | 1146 |