Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107028.1 | NP_001100498.1 | 5 | 523 | ||
XM_006246996.2 | XP_006247058.1 | 5 | 520 |