Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271003.1 | NP_001257932.1 | 4 | 834 | ||
NM_001271004.1 | NP_001257933.1 | 4 | 791 | |||
NM_006528.3 | NP_006519.1 | 4 | 867 | |||
GenBank mRNA | AK024817.1 | - | 4 | 629 | ||
AK075425.1 | - | 4 | 552 | |||
AK075427.1 | - | 4 | 679 | |||
AK092499.1 | - | 4 | 596 | |||
AK129833.1 | - | 4 | 629 | |||
AK313260.1 | - | 4 | 629 | |||
AY691946.1 | - | 4 | 605 | |||
BC005330.1 | - | 4 | 629 | 3929933 | ||
BC034155.1 | - | 3 | 352 | |||
D29992.1 | - | 4 | 610 | |||
DA833200.1 | - | 4 | 552 | |||
L27624.1 | - | 4 | 592 |