Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044238.1 | NP_001037703.1 | 2 | 250 | ||
GenBank mRNA | BC105620.1 | - | 2 | 250 | 7453591 |