Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 12, 13 | 4574 | ||
GenBank mRNA | AK123596.1 | - | 11, 12 | 2773 | ||
AK160365.1 | - | 12, 13 | 4630 | |||
BC010046.2 | - | 12, 13 | 1519 | |||
BC011839.2 | - | 12, 13 | 1519 | |||
BC014555.2 | - | 12, 13 | 1519 | |||
BC136661.1 | - | 12, 13 | 4488 | 9020651 | ||
BX649178.1 | - | 11, 12 | 2459 | |||
U02082.1 | - | 10, 11 | 1315 |
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 11, 12 | 3492 | |||
GenBank mRNA | DQ786236.1 | - | 6, 7 | 629 |