Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024993.1 | NP_001020164.1 | 5 | 273 | ||
XM_008768051.1 | XP_008766273.1 | 5 | 615 | |||
GenBank mRNA | BC092210.1 | - | 5 | 273 | 7311688 | |
EU327027.1 | - | 5 | 278 | |||
EU327028.1 | - | 4 | 234 | |||
EU327029.1 | - | 5 | 331 | |||
EU327030.1 | - | 4 | 195 | |||
EU327031.1 | - | 3 | 182 | |||
FQ213000.1 | - | 5 | 261 |
Gene Symbol: LOC103694314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772543.1 | XP_008770765.1 | 1 | 2148 |