Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006765.3 | NP_006756.2 | 1, 2 | 487 | ||
NM_178234.2 | NP_839952.1 | 1, 2 | 487 | |||
GenBank mRNA | BC010370.1 | - | 1, 2 | 355 | 4334284 | |
BT020002.1 | - | 1, 2 | 143 | |||
DA796279.1 | - | 1, 2 | 487 | |||
U42349.1 | - | 1, 2 | 300 |