Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098788.1 | NP_001092258.1 | 2 | 213 | ||
XM_006247314.2 | XP_006247376.1 | 3 | 782 | |||
XM_006247315.2 | XP_006247377.1 | 3 | 771 |