Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006765.3 | NP_006756.2 | 4, 5 | 910 | ||
NM_178234.2 | NP_839952.1 | 4, 5 | 910 | |||
XM_011544651.2 | XP_011542953.1 | 3, 4 | 453 | |||
XM_011544652.2 | XP_011542954.1 | 3, 4 | 453 | |||
XM_011544654.2 | XP_011542956.1 | 3, 4 | 453 | |||
XM_017013860.1 | XP_016869349.1 | 3, 4 | 453 | |||
XM_017013861.1 | XP_016869350.1 | 3, 4 | 453 | |||
GenBank mRNA | BC010370.1 | - | 4, 5 | 778 | 4334284 | |
BT020002.1 | - | 4, 5 | 566 | |||
U42349.1 | - | 4, 5 | 723 |