Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750935.3 | XP_003750983.1 | 5 | 1473 | ||
XM_006247467.2 | XP_006247529.1 | 6 | 1479 |