Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC303566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017988.1 | NP_001017988.1 | 2 | 263 | ||
XM_006247329.2 | XP_006247391.1 | 3 | 336 | |||
GenBank mRNA | AY623030.1 | - | 2 | 263 |