Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC303566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017988.1 | NP_001017988.1 | 4 | 1483 | ||
XM_006247329.2 | XP_006247391.1 | 5 | 1556 | |||
GenBank mRNA | AY623029.1 | - | 1 | 34 | ||
AY623030.1 | - | 4 | 1483 |