Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039019.1 | NP_001034108.1 | 10 | 1459 | ||
XM_006247505.2 | XP_006247567.1 | 10 | 1467 | |||
XM_008768283.1 | XP_008766505.1 | 10 | 1468 | |||
GenBank mRNA | BC099126.1 | - | 10 | 1459 | 7455959 |