Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107056.2 | NP_001100526.1 | 7 | 1926 | ||
XM_008768357.1 | XP_008766579.1 | 6 | 1728 | |||
GenBank mRNA | BC169069.1 | - | 7 | 1926 | 5623837 |