Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182820.1 | NP_877972.1 | 7 | 658 | ||
XM_006247586.2 | XP_006247648.1 | 9 | 837 | |||
XM_006247588.2 | XP_006247650.1 | 9 | 836 | |||
XM_006247595.2 | XP_006247657.1 | 8 | 806 | |||
XM_008768359.1 | XP_008766581.1 | 9 | 837 | |||
XM_008768360.1 | XP_008766582.1 | 9 | 813 | |||
XM_008768361.1 | XP_008766583.1 | 7 | 749 | |||
XM_008768362.1 | XP_008766584.1 | 7 | 725 | |||
XM_008768363.1 | XP_008766585.1 | 8 | 807 | |||
XM_008768364.1 | XP_008766586.1 | 8 | 783 | |||
GenBank mRNA | AY327409.1 | - | 7 | 658 | ||
BC081911.1 | - | 8 | 742 | 7111907 |