Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005672.4 | NP_005663.2 | 3 | 995 | ||
NR_033343.1 | 3 | 1148 | ||||
GenBank mRNA | AF043498.1 | - | 3 | 959 | ||
AY358912.1 | - | 3 | 928 | |||
BC023582.2 | - | 3 | 945 | 4840974 | ||
BC048808.1 | - | 3 | 945 | |||
BC065183.1 | - | 3 | 945 | 5557096 |