Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107060.1 | NP_001100530.1 | 1 | 626 | ||
GenBank mRNA | BC091245.1 | - | 1 | 272 |