Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006981.3 | NP_008912.2 | 3, 4 | 1687 | ||
NM_173199.2 | NP_775291.1 | 3, 4 | 1687 | |||
NM_173200.2 | NP_775292.1 | 4, 5 | 1759 | |||
XM_017015162.1 | XP_016870651.1 | 2, 3 | 1619 | |||
GenBank mRNA | AK226108.1 | - | 3, 4 | 1669 | ||
D78579.1 | - | 3, 4 | 1689 | |||
D85242.1 | - | 1, 2 | 58 | |||
S81243.1 | - | 3, 4 | 1574 | |||
U12767.1 | - | 2, 3 | 1050 | |||
X89894.1 | - | 3, 4 | 1645 |