Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107078.2 | NP_001100548.1 | 2 | 359 | ||
XM_006248693.2 | XP_006248755.1 | 3 | 732 | |||
XM_006248694.2 | XP_006248756.1 | 3 | 552 | |||
XM_006248695.2 | XP_006248757.1 | 3 | 729 | |||
XM_006248696.2 | XP_006248758.1 | 2 | 587 | |||
XM_006248697.2 | XP_006248759.1 | 2 | 584 | |||
XM_006248698.2 | XP_006248760.1 | 2 | 436 | |||
GenBank mRNA | BC169079.1 | - | 2 | 359 | 7937326 | |
FQ214252.1 | - | 2 | 553 |