Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270598.1 | NP_001257527.1 | 6 | 703 | ||
NM_001270599.1 | NP_001257528.1 | 6 | 703 | |||
GenBank mRNA | FQ222567.1 | - | 6 | 471 |