Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005436.4 | NP_005427.2 | 6 | 1223 | ||
GenBank mRNA | AK292593.1 | - | 6 | 1223 | ||
BC036757.2 | - | 6 | 1062 | 5549623 | ||
BC064391.1 | - | 6 | 1049 | 5745698 | ||
S72869.1 | - | 6 | 1027 |