Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100556.1 | NP_001094026.1 | 2 | 461 | ||
XM_006248398.2 | XP_006248460.1 | 2 | 791 | |||
XM_008768731.1 | XP_008766953.1 | 2 | 1313 | |||
GenBank mRNA | DY311373.1 | - | 2 | 496 |