Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123352.1 | NP_001116824.1 | 2 | 233 | ||
XM_008768732.1 | XP_008766954.1 | 2 | 971 | |||
GenBank mRNA | BC088235.1 | - | 2 | 226 | ||
BC161919.1 | - | 2 | 233 | 8369987 |
Gene Symbol: LOC103693563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768673.1 | XP_008766895.1 | 2 | 363 | ||
XM_008768674.1 | XP_008766896.1 | 2 | 970 | |||
XM_008768675.1 | XP_008766897.1 | 2 | 399 |