Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000496.1 | NP_001000496.1 | 1 | 798 |
Gene Symbol: Olr1543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001104.1 | NP_001001104.1 | 1 | 798 |