Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768605.1 | XP_008766827.1 | 1 | 728 |
Gene Symbol: Bach1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107113.1 | NP_001100583.1 | 4 | 1742 | ||
XM_003751009.3 | XP_003751057.2 | 4 | 1787 | |||
XM_006248018.2 | XP_006248080.1 | 4 | 1787 | |||
XM_006248019.2 | XP_006248081.1 | 4 | 2946 | |||
XM_006248020.2 | XP_006248082.1 | 4 | 2803 | |||
XM_008768581.1 | XP_008766803.1 | 4 | 3546 |