Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003478.3 | NP_003469.2 | 6 | 1287 | ||
XM_005271682.2 | XP_005271739.1 | 5 | 894 | |||
XM_011543013.1 | XP_011541315.1 | 6 | 1071 | |||
XM_017018363.1 | XP_016873852.1 | 6 | 1071 | |||
XM_017018364.1 | XP_016873853.1 | 6 | 632 | |||
XM_017018365.1 | XP_016873854.1 | 5 | 596 | |||
GenBank mRNA | AF017061.1 | - | 6 | 623 | ||
AF327710.1 | - | 6 | 620 | |||
AK125294.1 | - | 6 | 1236 | |||
AK292575.1 | - | 6 | 1050 | |||
BC063306.1 | - | 6 | 1050 | 30331132 | ||
X81882.1 | - | 6 | 709 |