Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020638.2 | NP_065689.1 | 1 | 217 | ||
GenBank mRNA | AB037973.1 | - | 1 | 71 | ||
AB047858.1 | - | 1 | 203 | |||
AF263537.1 | - | 1 | 217 | |||
AY358323.1 | - | 1 | 104 | |||
BC069333.1 | - | 1 | 145 | 7262193 | ||
BC096713.1 | - | 1 | 110 | 40000232 | ||
BC098147.1 | - | 1 | 110 | 40000231 | ||
BC098252.1 | - | 1 | 110 | 40000230 |