Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047092.1 | NP_001040557.1 | 6 | 1188 | ||
XM_006248930.2 | XP_006248992.1 | 9 | 1724 | |||
XM_006248931.2 | XP_006248993.1 | 8 | 1611 | |||
XM_006248932.2 | XP_006248994.1 | 7 | 2021 | |||
XM_006248933.2 | XP_006248995.1 | 7 | 1938 | |||
XM_008768970.1 | XP_008767192.1 | 7 | 1726 | |||
GenBank mRNA | AJ879913.1 | - | 6 | 1188 | ||
BC160815.1 | - | 6 | 1207 | 7365526 |