Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107128.2 | NP_001100598.2 | 6 | 898 | ||
NM_001142945.1 | NP_001136417.1 | 5 | 802 | |||
XM_008768974.1 | XP_008767196.1 | 3 | 1267 | |||
XM_008768975.1 | XP_008767197.1 | 6 | 956 |