Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107138.1 | NP_001100608.1 | 4 | 390 | ||
XM_006249300.2 | XP_006249362.1 | 4 | 967 | |||
XM_006249301.2 | XP_006249363.1 | 4 | 964 | |||
XM_008769224.1 | XP_008767446.1 | 3 | 862 | |||
XM_008769225.1 | XP_008767447.1 | 3 | 859 |